Skip to Content
Merck
All Photos(1)

Key Documents

861448

Sigma-Aldrich

8-Azaguanine

98%

Synonym(s):

2-Amino-6-hydroxy-8-azapurine, 2-Amino-6-oxy-8-azapurine, Guanazolo

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N6O
CAS Number:
Molecular Weight:
152.11
Beilstein:
171098
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

98%

mp

>300 °C (lit.)

SMILES string

NC1=Nc2[nH]nnc2C(=O)N1

InChI

1S/C4H4N6O/c5-4-6-2-1(3(11)7-4)8-10-9-2/h(H4,5,6,7,8,9,10,11)

InChI key

LPXQRXLUHJKZIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F R Ridzlan et al.
Tropical biomedicine, 27(3), 632-638 (2011-03-15)
Leptospirosis is recognized as one of the important zoonotic diseases in the world including Malaysia. A total of 145 soil and water samples were collected from selected National Service Training Centres (NSTC) in Kelantan and Terengganu. The samples were inoculated
Y Xiao et al.
Hunan yi ke da xue xue bao = Hunan yike daxue xuebao = Bulletin of Hunan Medical University, 22(2), 113-115 (1997-01-01)
A human lung cancer cell line (A549) mutant with hypoxanthine guanine phosphoribosyltransferase (HGPRT) deficiency and resistance to 8-AG was established by treatment of the A549 cells with 3 micrograms.ml-1 N-methyl-N'-nitro-nitrosoguanidine (MNNG), and prescreened in 0.25% agarose DMEM semisolid medium containing
O N Serova et al.
Neuroscience and behavioral physiology, 26(5), 477-481 (1996-09-01)
The investigation was devoted to the role of the synthesis of protein and peptide factors during the formation of chemosensory memory in rats. Two models of gustatory memorization were used: conditioned taste aversion (CTA), induced by the association of the
Bashir Mir et al.
Nucleic acids research, 32(3), e25-e25 (2004-02-13)
The use of primary somatic cells in nuclear transfer procedure has opened a new opportunity to manipulate domestic animal genomes via homologous recombination. To date, while a few loci have been targeted in somatic cells using similar enrichment strategies as
Christian Pfeifle et al.
Anticancer research, 29(11), 4489-4496 (2009-12-25)
Despite improvements in the treatment of patients with Ewing family tumours (EFT) during the past decades, the prognosis for patients with advanced disease is still unsatisfying. New treatment strategies have to be developed. A hypoxanthine/aminopterin/thymidine (HAT)-sensitive EFT cell line was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service