Skip to Content
Merck
All Photos(1)

Key Documents

851787

Sigma-Aldrich

Stachyose hydrate

98%

Synonym(s):

α-D-Gal-(1→6)-α-D-Gal-(1→6)-α-D-Glc-(1→2)-β-D-Fru, β-D-Fructofuranosyl-O-α-D-galactopyranosyl-(1→6)-O-α-D-galactopyranosyl-(1→6)-α-D-glucopyranoside, Lupeose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O21 · xH2O
CAS Number:
Molecular Weight:
666.58 (anhydrous basis)
Beilstein:
5710666
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

optical activity

[α]24/D +133°, c = 4 in H2O

SMILES string

O.OC[C@H]1O[C@H](OC[C@H]2O[C@H](OC[C@H]3O[C@H](O[C@]4(CO)O[C@H](CO)[C@@H](O)[C@@H]4O)[C@H](O)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C24H42O21.H2O/c25-1-6-10(28)14(32)17(35)21(41-6)39-3-8-11(29)15(33)18(36)22(42-8)40-4-9-12(30)16(34)19(37)23(43-9)45-24(5-27)20(38)13(31)7(2-26)44-24;/h6-23,25-38H,1-5H2;1H2/t6-,7-,8-,9-,10+,11+,12-,13-,14+,15+,16+,17-,18-,19-,20+,21+,22+,23-,24+;/m1./s1

InChI key

YDBMRUQRXAFOAH-KTDNCYJLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Stachyose hydrate has been used as an analytical standard in identifying the components of various plant extracts.[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wheat and the irritable bowel syndrome?FODMAP levels of modern and ancient species and their retention during bread making.
Ziegler J U, et al.
Journal of functional foods, 25, 257-266 (2016)
Bioassay-guided fractionation of Artocarpus heterophyllus L. J33 variety fruit waste extract and identification of its antioxidant constituents by TOF-LCMS.
Daud M N H, et al.
Food Chemistry (2018)
J W Ren et al.
Phytotherapy research : PTR, 26(10), 1547-1554 (2012-02-24)
Chinese herbal medicine has long been used as a treatment for wounds. However, the underlying cellular and molecular mechanisms remain largely unknown. In this study it was shown that the proliferation of keratinocytes, which is known to play an important
Robert Turgeon et al.
Protoplasma, 248(1), 173-180 (2010-11-17)
Phloem loading is the process by which photoassimilates synthesized in the mesophyll cells of leaves enter the sieve elements and companion cells of minor veins in preparation for long distance transport to sink organs. Three loading strategies have been described:
Rafael Opazo et al.
PloS one, 7(9), e44783-e44783 (2012-09-18)
Soybean meal (SBM) is an important protein source in animal feed. However, the levels of SBM inclusion are restricted in some animal species by the presence of antinutritional factors (ANFs), including non-starch polysaccharides (NSPs) and α-galactosides (GOSs). The aim of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service