Skip to Content
Merck
All Photos(1)

Key Documents

84570

Sigma-Aldrich

Sarcosine methyl ester hydrochloride

≥97.0% (T)

Synonym(s):

N-Methylglycine methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$468.00

$468.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
50 G
$468.00

About This Item

Linear Formula:
CH3NHCH2COOCH3 · HCl
CAS Number:
Molecular Weight:
139.58
Beilstein:
3619268
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$468.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (T)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

impurities

≤3% sarcosine hydrochloride

mp

117-119 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.CNCC(=O)OC

InChI

1S/C4H9NO2.ClH/c1-5-3-4(6)7-2;/h5H,3H2,1-2H3;1H

InChI key

HQZMRJBVCVYVQA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D O H Teare et al.
The journal of physical chemistry. B, 109(44), 20923-20928 (2006-07-21)
A new class of protein-resistant film based on N-substituted glycine derivatives is described. Pulsed plasma deposited poly(N-acryloylsarcosine methyl ester) coatings are shown to be resistant toward the adsorption of fibrinogen and lysozyme. Deposition and UV irradiation of the polymer through
M A Lindsay et al.
Bioscience reports, 10(6), 557-561 (1990-12-01)
Ca(2+)-Induced insulin release from electropermeabilised islets is inhibited by the transglutaminase inhibitors monodansylcadaverine, glycine methylester, methylamine and cystamine but not by the control compounds dimethyl monodansylcadaverine and sarcosine methylester which lack the primary amine group. Neither monodansylcadaverine nor glycine methylester
C Alarcon et al.
Bioscience reports, 5(7), 581-587 (1985-07-01)
Glycine methyl ester, an inhibitor of transglutaminase, decreased glucose-stimulated insulin release and delayed proinsulin conversion in rat pancreatic islets pulse-labelled with L-[4-3H]phenylalanine. Sarcosine methyl ester, which does not inhibit transglutaminase activity, failed to affect insulin release and proinsulin conversion. The
A M Bersten et al.
Biochimica et biophysica acta, 762(3), 429-436 (1983-06-02)
The effects of competitive inhibitors of transglutaminase on the formation of myotubes by the fusion of myoblasts in vitro has been investigated. Myotube formation was inhibited when myoblasts from 11-day-old chick embryos were cultured in vitro in the presence of
A Sener et al.
Endocrinology, 117(1), 237-242 (1985-07-01)
The Ca2+-responsive enzyme transglutaminase, which catalyzes the cross-bridging of proteins, is present in pancreatic islet cells, but its participation in the process of insulin release remains to be documented. Glycine methylester (1.0-10.0 mM) inhibited, in a dose-related manner, transglutaminase activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service