Skip to Content
Merck
All Photos(1)

Key Documents

255084

Sigma-Aldrich

Sarcosine ethyl ester hydrochloride

99%

Synonym(s):

Ethyl sarcosinate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$98.50

$98.50


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$98.50

About This Item

Linear Formula:
CH3NHCH2COOC2H5 · HCl
CAS Number:
Molecular Weight:
153.61
Beilstein:
3911182
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$98.50


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

99%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

127-128 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].CCOC(=O)CNC

InChI

1S/C5H11NO2.ClH/c1-3-8-5(7)4-6-2;/h6H,3-4H2,1-2H3;1H

InChI key

NIDZUMSLERGAON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joan Carles Nadal et al.
Journal of chromatography. A, 1626, 461348-461348 (2020-08-17)
Solid-phase extraction (SPE) is a widely-used and very well-established sample preparation technique for liquid samples. An area of on-going focus for innovation in this field concerns the development of new and improved SPE sorbents that can enhance the sensitivity and/or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service