Skip to Content
Merck
All Photos(1)

Key Documents

81273

Sigma-Aldrich

Poly(L-lactide)

viscosity ~2.0 dL/g, 0.1 % (w/v) in chloroform(25 °C)

Synonym(s):

L-Lactide polymer, PLA, Poly(L-Lactic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$335.00

$335.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$335.00

About This Item

Linear Formula:
CH3O(C6H8O4)nH
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$335.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

ester terminated

Quality Level

form

solid

mol wt

Mw ~260,000

degradation timeframe

>3 years

impurities

≤0.5% water

ign. residue

≤0.1%

viscosity

~2.0 dL/g, 0.1 % (w/v) in chloroform(25 °C)

Mw/Mn

~1.5

Looking for similar products? Visit Product Comparison Guide

Application

medical device

Other Notes

end group: decyl ester

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Stoiber et al.
Journal of the mechanical behavior of biomedical materials, 104, 103652-103652 (2020-03-17)
For the proper function of small diameter vascular grafts their mechanical properties are essential. A variety of testing methods and protocols exists to measure tensile strength, compliance and viscoelastic material behavior. In this study the impact of the measurement protocol

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

Immunosuppressive tumor-associated myeloid cells (TAMC) are responsible for glioblastoma (GBM) resistance to immunotherapies and existing standard of care treatments. This mini-review highlights recent progress in implementing nanotechnology in advancing TAMC-targeted therapies for GBM.

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Questions

  1. Hello! What is the molecular weight of this product?

    1 answer
    1. As listed in the 'PROPERTIES' section, the molecular weight is~260,000 Da.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service