Skip to Content
Merck
Dimethyl carbonate analytical standard

Dimethyl carbonate

Dimethyl carbonate analytical standard
Synonym(s):
Carbonic acid dimethyl ester, Dimethyl carbonate
Linear Formula:
(CH3O)2CO
CAS Number:
Molecular Weight:
90.08
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing