Skip to Content
Merck
All Photos(1)

Key Documents

808822

Sigma-Aldrich

Tungsten diselenide

Crystal, 99.995%

Synonym(s):

Tungsten (IV) selenide, Tungsten selenide

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,700.00

$1,700.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,700.00

About This Item

Linear Formula:
WSe2
CAS Number:
Molecular Weight:
341.76
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

$1,700.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

description

Electrical properties: Semiconductor, p-type

Assay

99.995%

form

crystals

L × W × H

0.5 cm × 0.5 cm × 0.1-0.15 mm, typical size, some variation in dimensions may occur with different batches

primary crystallite size

~10 mm (hexagonal)

SMILES string

[Se]=[W]=[Se]

InChI

1S/2Se.W

InChI key

ROUIDRHELGULJS-UHFFFAOYSA-N

Related Categories

Application

The two dimensional layered inorganic nanomaterials have drawn significant attention recently owing to their thickness dependent physical and chemical properties. They find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc. Transition metal dischalcogenides represent an important class of inorganic 2D materials having the aforementioned applications.[1][2][3]

Preparation Note

Store under nitrogen atmosphere.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Biccai et al.
ACS nano, 13(6), 6845-6855 (2019-06-15)
Nanocomposite strain sensors, particularly those consisting of polymer-graphene composites, are increasingly common and are of great interest in the area of wearable sensors. In such sensors, application of strain yields an increase in resistance due to the effect of deformation
Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar JL, et al.
The Journal of Physical Chemistry C, 118, 3847-3863 (2014)
The super materials that could trump graphene.
Elizabeth Gibney
Nature, 522(7556), 274-276 (2015-06-19)
Sheneve Z Butler et al.
ACS nano, 7(4), 2898-2926 (2013-03-08)
Graphene's success has shown that it is possible to create stable, single and few-atom-thick layers of van der Waals materials, and also that these materials can exhibit fascinating and technologically useful properties. Here we review the state-of-the-art of 2D materials

Articles

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

Advances in scalable synthesis and processing of two-dimensional materials

For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service