Skip to Content
Merck
All Photos(2)

Key Documents

803855

Sigma-Aldrich

Schreiner′s Catalyst

greener alternative

Synonym(s):

1,3-Bis[3,5-bis(trifluoromethyl)phenyl]urea, N,N′-bis[3,5-bis(trifluoromethyl)phenyl]-Urea

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$191.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$191.00

About This Item

Empirical Formula (Hill Notation):
C17H8F12N2O
CAS Number:
Molecular Weight:
484.24
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

Quality Level

form

powder

reaction suitability

reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1)NC2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2

InChI

1S/C17H8F12N2O/c18-14(19,20)7-1-8(15(21,22)23)4-11(3-7)30-13(32)31-12-5-9(16(24,25)26)2-10(6-12)17(27,28)29/h1-6H,(H2,30,31,32)

InChI key

YGCOMBKZFUMALE-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Schreiner′s catalyst is commonly used as a hydrogen-bond donor in the activation of carbonyls, nitroolefins, imines etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bonding Organocatalysed Friedel-Crafts Alkylation of Aromatic and Heteroaromatic Systems with Nitroolefins
Dessole G,et al.
Synlett, 13, 2374-2378 (2004)
Sarah A Kavanagh et al.
Organic & biomolecular chemistry, 6(8), 1339-1343 (2008-04-04)
N,N'-Diarylureas have been shown to efficiently catalyse sulfonium ylide-mediated aldehyde epoxidation reactions for the first time. These processes are of broad scope and can be coupled with a subsequent Cu(II) ion-catalysed Meinwald rearrangement to give an efficient and convenient protocol
Sonia S So et al.
Organic letters, 13(4), 716-719 (2011-01-14)
Boronate ureas are introduced as a new class of noncovalent catalysts for conjugate addition reactions with enhanced activity. Through intramolecular coordination of the urea functionality to a strategically placed Lewis acid, rate enhancements up to 10 times that of more

Related Content

Boronate ureas benefit from internal Lewis acid coordination of the urea cabonyl oxygen and the strategically placed boron. As a result of this structural feature, boronate ureas can be rendered more acidic than conventional urea hydrogen bond donor catalysts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service