Skip to Content
Merck
All Photos(1)

Key Documents

803316

Sigma-Aldrich

Sulfo-LC-SPDP (sulfosuccinimidyl 6-[3′-(2-pyridyldithio)propionamido]hexanoate)

Synonym(s):

2,5-Dioxo-3-sulfo-1-pyrrolidinyl 6-[[1-oxo-3-(2-pyridinyldithio)propyl]amino]hexanoate, Sulfo-LC SPDP, Sulfosuccinimidyl 6-(3′-(2-pyridyldithio)propionamido)hexanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N3NaO8S3
Molecular Weight:
473.48
MDL number:
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥90%

Quality Level

form

powder

mol wt

527.57

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

soluble (>95 mg/ml in water)

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1CC(S(=O)([O-])=O)C(N1OC(CCCCCNC(CCSSC2=CC=CC=N2)=O)=O)=O.[Na+]

InChI

1S/C18H23N3O8S3.Na/c22-14(8-11-30-31-15-6-3-5-10-20-15)19-9-4-1-2-7-17(24)29-21-16(23)12-13(18(21)25)32(26,27)28;/h3,5-6,10,13H,1-2,4,7-9,11-12H2,(H,19,22)(H,26,27,28);/q;+1/p-1

InChI key

ZAPNXDUFCQIHFS-UHFFFAOYSA-M

General description

The SPDP reagents are a unique group of amine- and sulfhydryl-reactive heterobifunctional crosslinkers. Whether they are used to form amine-to-amine or amine-to-sulfhydryl crosslinks among molecules, the SPDP reagents produce disulfide-containing linkages that can be cleaved later with reducing agents such as dithiothreitol (DTT).

Features and Benefits

  • Reactive groups: NHS ester and pyridyldithiol
  • Reactive towards: amino and sulfhydryl groups
  • Releases a detectable by-product when reacted with a free sulfhydryl group; by measuring the release of pyridine-2-thione at 343 nm, the reaction can be easily followed
  • Water soluble, sulfo-NHS ester reacts rapidly with any primary amine-containing molecule
  • Disulfide bond in the spacer arm is readily cleaved by 10-50 mM DTT or TCEP at pH 8.5
  • Spacer arm is also easily cleaved using reducing SDS-PAGE sample loading buffer
  • Cleavable crosslinker allows separation of crosslinked products
  • Sulfo-LC-SPDP crosslinker is not membrane permeable, allowing cell surface labeling
  • Compare to other varieties of SPDP-type reagents, including pegylated forms

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preparation of antibody-toxin conjugates.
A J Cumber et al.
Methods in enzymology, 112, 207-225 (1985-01-01)
Neurath, A. R. and Strick, N.
J. Virol. Methods, 3, 155-165 (1981)
J Carlsson et al.
The Biochemical journal, 173(3), 723-737 (1978-09-01)
A heterobifunctional reagent, N-succinimidyl 3-(2-pyridyldithio)propionate, was synthesized. Its N-hydroxysuccinimide ester group reacts with amino groups and the 2-pyridyl disulphide structure reacts with aliphatic thiols. A new thiolation procedure for proteins is based on this reagent. The procedure involves two steps.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service