Skip to Content
Merck
All Photos(1)

Key Documents

QBD10374

Sigma-Aldrich

SPDP-dPEG®4-NHS ester

Synonym(s):

2,5-Dioxo-1-pyrrolidinyl 17-oxo-19-(2-pyridinyldithio)-4,7,10,13-tetraoxa-16-azanonadecanoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$418.00

$418.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$418.00

About This Item

Empirical Formula (Hill Notation):
C23H33N3O9S2
CAS Number:
Molecular Weight:
559.65
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$418.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: thiol reactive
reagent type: cross-linking reagent
reactivity: amine reactive

functional group

NHS ester

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(NCCOCCOCCOCCOCCC(ON1C(CCC1=O)=O)=O)CCSSC2=NC=CC=C2

InChI

1S/C23H33N3O9S2/c27-19(7-18-36-37-20-3-1-2-8-25-20)24-9-11-32-13-15-34-17-16-33-14-12-31-10-6-23(30)35-26-21(28)4-5-22(26)29/h1-3,8H,4-7,9-18H2,(H,24,27)

InChI key

QTVBGVZVUCIFAH-UHFFFAOYSA-N

Features and Benefits

SPDP-dPEG®4-NHS ester has an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a sulfhydryl-reactive 2-pyridyldithiol group at the opposite end. This has a single molecular weight PEG spacer arm, which confers greater water solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Protein aggregation and precipitation also are greatly reduced with SPDP-dPEG crosslinkers compared to crosslinkers that have only hydrocarbon spacers. dPEG spacer of specific length (23.1 Å) provides precise distance control between the SPDP moiety and the target molecule. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using SPDP reagents.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service