803049
Folic acid-(glutamic acid-13C5)
≥99 atom % 13C, ≥95% (CP)
Synonym(s):
Vitamin B9-(glutamic acid-13C5)
Select a Size
Select a Size
About This Item
Recommended Products
isotopic purity
≥99 atom % 13C
Quality Level
Assay
≥95% (CP)
form
powder
technique(s)
mass spectrometry (MS): suitable
color
white to light yellow
mp
>285 °C
application(s)
detection
mass shift
M+5
storage temp.
−20°C
SMILES string
O=C1C2=C(N=CC(CNC3=CC=C(C(N[13CH]([13C](O)=O)[13CH2][13CH2][13C](O)=O)=O)C=C3)=N2)N=C(N)N1
InChI
1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/i5+1,6+1,12+1,13+1,18+1
InChI key
OVBPIULPVIDEAO-BCTWCYHZSA-N
General description
Folic acid-(glutamic acid-13C5) is an isotope of vitamin B9 wherein all the five glutamate carbons are replaced by 13C6 carbon.
Application
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service