Skip to Content
Merck
All Photos(1)

Key Documents

607835

Sigma-Aldrich

L-Aspartic acid-13C4,15N

98 atom % 13C, 98 atom % 15N, 95% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$2,070.00

$2,070.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$2,070.00

About This Item

Linear Formula:
HO213C13CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
138.07
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$2,070.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 13C
98 atom % 15N

Quality Level

Assay

95% (CP)

form

solid

optical activity

[α]25/D +25.0°, c = 2 in 5 M HCl

technique(s)

bio NMR: suitable

mp

>300 °C (dec.) (lit.)

mass shift

M+5

SMILES string

[15NH2][13C@@H]([13CH2][13C](O)=O)[13C](O)=O

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1/i1+1,2+1,3+1,4+1,5+1

InChI key

CKLJMWTZIZZHCS-NYTNKSBQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service