Skip to Content
Merck
All Photos(1)

Key Documents

802557

Sigma-Aldrich

Aquivion® D98-25BS

greener alternative

liquid, dispersion, 25% in water, PFSA eq. wt. 980 g/mole SO3H, contains CF3 polymer chain ends as stabilizer

Synonym(s):

Aquivion® SO3H-form, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$386.00
250 ML
$2,590.00

$386.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$386.00
250 ML
$2,590.00

About This Item

Linear Formula:
(C4HF7O4S .C2F4)x
UNSPSC Code:
26111700
NACRES:
NA.23

$386.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

dispersion
liquid

eq. wt.

980±40g/mole (SO3H)

contains

CF3 polymer chain ends as stabilizer

composition

25% in water

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

25±1 % (w/w)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Aquivion® PFSA dispersions are derived from a short-side-chain (SSC) copolymer of Tetrafluoroethylene and Sulfonyl Fluoride Vinyl Ether (SFVE), CF2=CF2-O-(CF2)2-SO2F, industrially produced by Syensqo Group. The resulting perfluoropolymer features functional groups in their sulfonic acid form (SO3H). Aquivion® D98-25BS is a perfluorosulfonic acid (PFSA) ionomer dispersion with an Equivalent Weight (EW) of 980 g/eq. The EW of an ionomer represents the weight of the polymer required to provide 1 mole of exchangeable protons, inversely related to the ion exchange capacity (IEC). This dispersion is chemically stabilized, with a significant proportion of polymer end groups converted to an inert trifluoromethyl (CF3) configuration, enhancing the polymer′s lifetime. Aquivion® D98-25BS contains a polymer concentration of 25 wt% in a solvent system that is over 99% water and free of ethers.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for use as Proton Exchange Membrane (PEM) Fuel Cells. Click here for more information.

Application

The superacidity of Aquivion® PFSA resins, combined with their high functional group concentration and electrochemical stability, makes them a critical ingredient in heterogeneous catalysts. Due to their excellent proton conductivity at low water uptake, these resins are frequently employed in research as ionomer binders within the catalyst layers of proton exchange membranes (PEM) for fuel cells and water electrolyzers. Additionally, they can be used in fabricating separator membranes (PEMs) either as stand-alone materials or reinforced with PTFE and are sometimes added to the surfaces of specialty fluoropolymers to enhance wettability. Aquivion® D98-25BS, with its short side chain and high equivalent weight, is a promising ionomer for balancing catalyst activity and stability. Ionomers with higher equivalent weights, such as D98-25BS, exhibit greater durability in long-term electrochemical stability tests. Moreover, D98-25BS ionomer catalysts can form smaller aggregates and agglomerates in certain catalyst systems. In these instances, the enhanced interaction between the catalyst and the D98-25BS binder can improve reaction kinetics, reduce overpotential, and enhance catalyst utilization compared to lower equivalent weight ionomers.

Features and Benefits

  • Very high concentration of functional groups for strong proton (or Na+, K+, Li+, etc.) conductivity
  • High softening point for high temperature operation well above 80 ºC
  • Strong water retention for better fuel cell operation at low-humidity conditions for membranes or catalyst inks that are utilizing this dispersion material

Legal Information

Aquivion is a registered trademark of Syensqo Group

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Are Nafion 117 containing solution and Aquivion D98-25BS are same?

    1 answer
    1. Nafion 117 and Aquivion D98-25BS are not identical. Various literature sources suggest that the major differences between Nafion and Aquivion are the different side chain lengths. However, Aquivion D98-25BS is considered to be a good alternative for Nafion™ 117.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service