B10900
2,1,3-Benzothiadiazole
98%
Synonym(s):
Piazthiole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4N2S
CAS Number:
Molecular Weight:
136.17
Beilstein:
112408
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
crystals
bp
206 °C (lit.)
mp
42-44 °C (lit.)
SMILES string
c1ccc2nsnc2c1
InChI
1S/C6H4N2S/c1-2-4-6-5(3-1)7-9-8-6/h1-4H
InChI key
PDQRQJVPEFGVRK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
203.0 °F - closed cup
Flash Point(C)
95 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhixiong Cai et al.
Nanotechnology, 28(42), 425102-425102 (2017-08-03)
Nanoplatform integrated with photothermal therapy (PTT) and chemotherapy has been recognized a promising agent for enhancing cancer therapeutic outcomes, but still suffer from less controllability for optimizing their synergistic effects. We fabricated glutathione (GSH) responsive micelles incorporated with semiconducting polymer
Thiago O Carvalho et al.
The Journal of organic chemistry, 84(9), 5118-5128 (2019-04-09)
A water-soluble and charge-tagged palladium complex (PdMAI) was found to function inside breast cancer live cells of the MCF-7 lineage as an efficient catalyst for cross-coupling reaction. PdMAI, bearing two ionophilic task-specific ionic liquids as ligands, efficiently catalyzed both in
Gözde Murat Saltan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 188, 372-381 (2017-08-02)
In an approach to develop efficient organic optoelectronic devices to be used in light-driven systems, a series of three thiophene linked benzimidazole conjugates were synthesized and characterized. The combination of two thiophene rings to a benzimidazole core decorated with different
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service