Skip to Content
Merck
All Photos(1)

Key Documents

798088

Sigma-Aldrich

Fluorescent nanodiamond

Nitrogen vacancy ~3 ppm NV centers, 120 nm avg. part. size, 1 mg/mL in deionized water

Synonym(s):

Diamond nanopowder, Nanodiamonds

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$542.00

$542.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$542.00

About This Item

Linear Formula:
C
UNSPSC Code:
12352302
NACRES:
NA.23

$542.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

suspension

Quality Level

composition

Nitrogen vacancy, ~3 ppm NV centers

concentration

1 mg/mL in deionized water

avg. part. size

120 nm

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescent nanodiamonds (FNDs) can be prepared by subjecting synthetic diamond nanocrystallites to helium ions.[1] FND is biocompatible and its surface can be easily functionalized. It is a very useful tool in bioimaging mainly because the photons emitted from the defect centers of FND upon laser excitation can easily penetrate the tissue. The emission from FNDs is extremely stable.[1] The average size of the FNDs is 120 nm.
In fluorescent nanodiamonds, the nitrogen vacancy centers in the diamond lattice provide the fluorescence; unlike the quantum dots or organic dyes, the color centers in these don′t photobleach or photoblink. These outstanding properties of fluorescent nanodiamonds along with their biocompatibility, large surface area etc. make them promising bioimaging probes.
Product can be functionalized with carboxylic acid, hydroxyl, ketones, ethers, and/or hydrogen.

Application

Potential uses of FNDs are as in vivo nanoprobe for bioimaging, cell labelling and cell tracking.[1][2]

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Exocytosis of Fluorescent Nanodiamond and Its Use as a Long-Term Cell Tracker
Fang CY, et al.
Small, 7(23), 3363-3370 (2011)
Nanodiamonds for optical bioimaging
Y. Y. Hui, C-L Cheng, and H-C Chang
Journal of Physics D: Applied Physics, 43(37), 374021-374021 (2010)
Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting
B-M Chang, H-S Lin, L-J Su, et al.
Advances in Functional Materials, 23(46), 5737-5745 (2013)
Mass production and dynamic imaging of fluorescent nanodiamonds
Chang YR, et al.
Nature Nanotechnology, 3(5), 284-288 (2008)

Articles

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Questions

1–2 of 2 Questions  
  1. I would like to see the fluorescence response of 798088

    1 answer
    1. Please see the image below to review the spectral data for this product.

      Helpful?

  2. 1. Is your inflorescence nanodiamond dyed?
    2. What are the excitation and emission wavelengths for them? are their quantum yield and brightness information available?
    3. What is the function of the PEG functionalization here? Does it reduce surface tension? Does the functional group affect the fluorescent intensity?
    4. Has it been used for some fluorescent tracing experiments?

    1 answer
    1. 1 The fluorescence originates from the nanodiamonds centers, such as nitrogen-vacancy (NV), NVN, or N3, and not from any fluorescent groups. Therefore, it is not dyed.
      2 The excitation/emission wavelengths are consistent for these products and are based on the nanodiamonds center. The catalog part numbers, center type, excitation peak, emission peak, and brightness information are as follows:
      900173, NVN, 480 nm, 520 nm (green), ~90x vs FITC
      All other products, NV, 570 nm, 680 nm (red), ~70x brighter vs Atto 647 Two important notes provided were:
      Green fluorescence is brighter than red.
      Longer wavelengths, such as red light, have better penetrability.
      3 PEG functionalization historically reduces non-specific binding and aggregation in buffered media. Carboxylated particles are commonly used for sensing and imaging applications, or to conduct further syntheses in the lab. However, in biological media/buffer (e.g., PBS), they are prone to aggregation, and surface coatings help to reduce this. Moreover, other functionalizations, such as biotin or antibodies, allow for selective targeting.
      4 Threr is information available that it has been used in fluorescent imaging experiments.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service