Skip to Content
Merck
All Photos(1)

Key Documents

791903

Sigma-Aldrich

Conductive silver printing ink, resistivity 30 - 35 μΩ-cm

Synonym(s):

Dyesol® DYAG350 conductive silver ink, Screen printable silver paste

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

description

volume resistivity 30 - 35 μΩ-cm at a 180°C cure temperature

form

paste (white)

composition

Solids content, 65-75%

viscosity

6000-9000 mPa.s (at shear rate of 10 sec-1 at 25°C)

General description

Conductive silver printing ink, resistivity 30-35 μΩ-cm, is a combination of polymer coated silver particles and silver-based compounds in an organic medium. It is a low cost conductive ink that can be printed on a variety of substrates like polyester, glass, and indium tin oxide (ITO) which can be cured at 150-180 °C.[1][2]

Application

Conductive silver inks can be coated in between reduced graphene oxide (rGO) and copper surfaces to stabilize the charge carrying medium which can be used in the fabrication of a thermistor.[3] Laser induced forward transfer (LIFT) may use conductive silver printing inks to print indium tin oxide nanoparticles based thin films for solar cells and organic light emitting diodes (OLEDS) based applications.[4]
This Conductive Silver ink is an economical grade of specially formulated, screen-printable, conductive silver ink which provides good conductivity at low cure temperatures. This conductive silver ink provides low resistance of printed conducting tracks.

This Conductive Silver Printing Ink is comprised of a mixture of pure silver particles and organometallic silver compound in an organic medium. When printed on polyester (PET or PEN) films and then cured at 150 - 180°C, the particles consolidate to form a chemically-welded, continuous silver track to yield low bulk resistivity.

Legal Information

Dyesol is a registered trademark of Greatcell Solar

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-based inkjet inks for printed electronics.
Kamyshny A, et al.
Open Applied Physics Journal, 4(1), 4153-4158 (2011)
Direct ink-jet printing and low temperature conversion of conductive silver patterns.
Smith PJ, et al.
J. Mater. Sci., 41(13), 4153-4158 (2006)
Design guidelines of laser reduced graphene oxide conformal thermistor for IoT applications.
Romero FJ, et al.
Sensors and actuators A, Physical, 274(1), 148-154 (2018)
Conductive silver ink printing through the laser-induced forward transfer technique.
Florian C, et al.
Applied Surface Science, 336(1), 304-308 (2015)

Articles

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

See All

Questions

  1. silver ink printed paper can use in oven?

    1 answer
    1. Unfortunately, there is no information on the curing process for this product 791903 when used on paper. Conductive Silver Printing Ink is ideal for use in plastic/printed electronics and can be used with conventional screen print technology and can be cured at 150-180 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service