Skip to Content
Merck
All Photos(1)

Key Documents

779873

Sigma-Aldrich

Methyl-β-cyclodextrin

Wacker Chemie AG, ≥98.0% cyclodextrin basis

Synonym(s):

Cavasol® W7 M Life Science, MβCD, Methyl-beta-cyclodextrin

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$1,390.00
500 G
$2,060.00
1 KG
$2,610.00
10 KG
$37,240.00

$1,390.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$1,390.00
500 G
$2,060.00
1 KG
$2,610.00
10 KG
$37,240.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

$1,390.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% cyclodextrin basis

form

powder

optical activity

[α]/D 154.0 to 156.0°

mol wt

Mw 1310 g/mol

manufacturer/tradename

Wacker Chemie AG

extent of labeling

1.7-1.9 molar substitution

impurities

≤0.10% unsubstituted cyclodextrin
≤0.20% chloride
≤0.50% reducing substances
≤50 ppm volatile organics

ign. residue

≤0.50%

loss

≤5.0% loss on drying

color

white to off-white

mp

180-182 °C (lit.)

absorption

≤0.10 at 350-600 nm in solution at 10%
≤1.00 at 220-350 nm in solution at 10%

SMILES string

O1C2OC(C(OC3OC(C(OC4OC(C(OC5OC(C(OC6OC(C(OC7OC(C(OC8OC(C1C(C8OC)O)COC)C(C7OC)O)COC)C(C6OC)O)COC)C(C5OC)O)COC)C(C4OC)O)COC)C(C3OC)O)COC)C(C2OC)O)COC

InChI

1S/C56H98O35/c1-64-15-22-36-29(57)43(71-8)50(78-22)86-37-23(16-65-2)80-52(45(73-10)30(37)58)88-39-25(18-67-4)82-54(47(75-12)32(39)60)90-41-27(20-69-6)84-56(49(77-14)34(41)62)91-42-28(21-70-7)83-55(48(76-13)35(42)63)89-40-26(19-68-5)81-53(46(74-11)33(40)61)87-38-24(17-66-3)79-51(85-36)44(72-9)31(38)59/h22-63H,15-21H2,1-14H3

InChI key

QGKBSGBYSPTPKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl-β-cyclodextrin is a macrocyclic compound that can form inclusion complexes with a number of guest molecules.[1] It shows higher solubility in aqueous solutions and greater solubilizing and complexing power compared to the parent β-cyclodextrin.[2] MβCD can be used to enhance the enzyme activity and enantioselectivity of subtilisin enzyme in organic solvents.[1]

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
For R&D Use

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

368.6 °F

Flash Point(C)

187 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and solid-state characterization of inclusion complexes formed between miconazole and methyl-?-cyclodextrin.
Ribeiro A
Aaps Pharmscitech, 9(4), 1102-1109 (2008)
Improved enzyme activity and enantioselectivity in organic solvents by methyl-?-cyclodextrin.
Griebenow K
Journal of the American Chemical Society, 121(36), 8157-8163 (1999)
Sara Rodríguez-Acebes et al.
The Biochemical journal, 420(2), 305-315 (2009-03-06)
Cholesterol homoeostasis is critical for cell viability and proliferation. The SREBP (sterol regulatory element-binding protein) pathway is crucial for the maintenance of cholesterol homoeostasis. This pathway is controlled by cholesterol and cholesterol-derived oxysterols. J774 cells cannot convert desmosterol into cholesterol
Mario Menschikowski et al.
Cancer cell international, 11, 4-4 (2011-02-16)
Increasing evidences show that beyond its role in coagulation, endothelial protein C receptor (EPCR) interferes with carcinogenesis. Pro-carcinogenic effects of EPCR were linked with a raised generation of activated protein C (aPC) and anti-apoptotic signalling. This study was carried out
Laurie Hamtiaux et al.
PloS one, 6(10), e26823-e26823 (2011-11-03)
The antitumoral properties of endocannabinoids received a particular attention these last few years. Indeed, these endogenous molecules have been reported to exert cytostatic, apoptotic and antiangiogenic effects in different tumor cell lines and tumor xenografts. Therefore, we investigated the cytotoxicity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service