Skip to Content
Merck
All Photos(1)

Key Documents

776823

Sigma-Aldrich

6-Hydroxybenzothiazole-2-carbonitrile

96%

Synonym(s):

2-cyano-6-hydroxybenzothiazole

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$121.00

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$121.00

About This Item

Empirical Formula (Hill Notation):
C8H4N2OS
CAS Number:
Molecular Weight:
176.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

211-213 °C

functional group

nitrile

storage temp.

2-8°C

SMILES string

OC1=CC(SC(C#N)=N2)=C2C=C1

InChI

1S/C8H4N2OS/c9-4-8-10-6-2-1-5(11)3-7(6)12-8/h1-3,11H

InChI key

SQAVNBZDECKYOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arkadiy A Bazhin et al.
Nature chemical biology, 16(12), 1385-1393 (2020-08-12)
Mitochondrial membrane potential (ΔΨm) is a universal selective indicator of mitochondrial function and is known to play a central role in many human pathologies, such as diabetes mellitus, cancer and Alzheimer's and Parkinson's diseases. Here, we report the design, synthesis
Ce Shi et al.
PloS one, 15(12), e0243747-e0243747 (2020-12-15)
Firefly luciferase-based ATP detection assays are frequently used as a sensitive, cost-efficient method for monitoring hygiene in many industrial settings. Solutions of detection reagent, containing a mixture of a substrate and luciferase enzyme that produces photons in the presence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service