Skip to Content
Merck
All Photos(1)

Key Documents

30095

Sigma-Aldrich

D-Cysteine

≥99% (RT), for peptide synthesis

Synonym(s):

(S)-2-Amino-3-mercaptopropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$104.00
1 G
$276.00

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$104.00
1 G
$276.00

About This Item

Empirical Formula (Hill Notation):
C3H7NO2S
CAS Number:
Molecular Weight:
121.16
Beilstein:
1721407
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

D-Cysteine, ≥99% (RT)

Quality Level

Assay

≥99% (RT)

form

powder or crystals

optical activity

[α]20/D −7.6±1.0°, c = 5% in 5 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

~230 °C (dec.)

application(s)

peptide synthesis

SMILES string

N[C@H](CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m1/s1

InChI key

XUJNEKJLAYXESH-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Cysteine has been used in the functionalization of QDs for applications in photocatalysis.[1]
It can also be used in the synthesis of:
  • cysteine-based chiral carbon dots (CDs)[2]
  • D-luciferin and aminoluciferin analogs[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Unnatural isomer of cysteine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wang et al.
Nanomaterials (Basel, Switzerland), 8(12) (2018-12-14)
Chiral recognition is of fundamental importance in chemistry and life sciences and the principle of chiral recognition is instructive in chiral separation and enantioselective catalysis. Non-chiral Ag nanoparticles (NPs) conjugated with chiral cysteine (Cys) molecules demonstrate strong circular dichroism (CD)
Angéla Takács et al.
Scientific reports, 10(1), 14287-14287 (2020-09-02)
Bortezomib (BOZ) is a proteasome inhibitor chemotherapeutic agent utilized to treat multiple myeloma and recently offered to cure melanoma. Bortezomib-induced neuropathy is one of the dose-limiting side-effects, which can be treated with antioxidants (e.g. alpha-lipoic acid-ALA and Vitamin B1-vit B1).
Yusuke Shibui et al.
Journal of toxicologic pathology, 30(3), 217-229 (2017-08-12)
Two 4-week repeated-dose toxicity studies were conducted to evaluate the potential toxicity of l-cysteine and d-cysteine. In one study, three groups of 6 male rats were each administered l-cysteine once daily by gavage at doses of 500, 1,000, or 2,000
Antonia Asimakopoulou et al.
British journal of pharmacology, 169(4), 922-932 (2013-03-16)
Hydrogen sulfide (H₂S) is a signalling molecule that belongs to the gasotransmitter family. Two major sources for endogenous enzymatic production of H₂S are cystathionine β synthase (CBS) and cystathionine γ lyase (CSE). In the present study, we examined the selectivity
Nar Singh Chauhan et al.
Indian journal of microbiology, 57(3), 299-306 (2017-09-15)
A metagenomic library of sea sediment metagenome containing 245,000 recombinant clones representing ~ 2.45 Gb of sea sediment microbial DNA was constructed. Two unique arsenic resistance clones, A7 and A12, were identified by selection on sodium arsenite containing medium. Clone A7 showed a

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service