776254
Nickel(II) bis(trifluoromethanesulfonimide) hydrate
95%
About This Item
Recommended Products
Quality Level
Assay
95%
form
solid
reaction suitability
core: nickel
reagent type: catalyst
mp
281-286 °C
storage temp.
2-8°C
SMILES string
[Ni](N([S](=O)(=O)C(F)(F)F)[S](=O)(=O)C(F)(F)F)N([S](=O)(=O)C(F)(F)F)[S](=O)(=O)C(F)(F)F.O
InChI
1S/2C2F6NO4S2.Ni.H2O/c2*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;;/h;;;1H2/q2*-1;+2;
InChI key
UXESDHJDHBRMLI-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service