Skip to Content
Merck
All Photos(3)

Key Documents

708801

Sigma-Aldrich

Iron(III) trifluoromethanesulfonate

greener alternative

90%

Synonym(s):

Fe(CF3SO3)3, Ferric triflate, Ferric trifluoromethanesulfonate, Fe(CF3SO3)3, Fe(OTf)3, Iron 1,1,1-trifluoromethanesulfonic acid, Iron(III) triflate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$84.10

$84.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$84.10

About This Item

Empirical Formula (Hill Notation):
C3F9FeO9S3
CAS Number:
Molecular Weight:
503.05
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$84.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

90%

form

powder

reaction suitability

core: iron
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

183 °C

greener alternative category

SMILES string

[Fe+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Fe/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

OSHOQERNFGVVRH-UHFFFAOYSA-K

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-Jie Song et al.
ChemSusChem, 8(24), 4250-4255 (2015-11-28)
Using a metal triflate and Pd/C as catalysts, alkanes were prepared from bioderived furans in a one-pot hydrodeoxygenation (HDO) process. During the reaction, the metal triflate plays a crucial role in the ring-opening HDO of furan compounds. The entire reaction
Yang Li et al.
Macromolecular bioscience, 20(11), e2000146-e2000146 (2020-06-23)
The conducting polymer polyethylenedioxythiophene doped with polystyrene sulfonate (PEDOT:PSS) has received great attention in the field of wearable bioelectronics due to its tunable high electrical conductivity, air stability, ease of processability, biocompatibility, and recently discovered self-healing ability. It has been
Imtiyaz Ahmad Wani et al.
Organic & biomolecular chemistry, 16(16), 2910-2922 (2018-04-06)
Direct and expedient access to densely substituted tetrahydrocarbazoles and tetrahydrocycloheptadiindoles bearing multiple contiguous stereocentres has been achieved via a two-fold divergent diastereoselective (dr up to >99 : 1) transformation of 2-vinylindoles. The high-yielding conversions (yield up to 87%) that are amenable for
Raffaele Cucciniello et al.
ChemSusChem, 9(23), 3272-3275 (2016-11-24)
The present work deals with the production of monoalkyl glyceryl ethers (MAGEs) through a new reaction pathway based on the reaction of glycidol and alcohols catalyzed by Lewis acid-based catalysts. Glycidol is quantitatively converted with high selectivity (99 %) into MAGEs

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service