776149
HDMA
99%
Synonym(s):
1-[(Dimethylamino)(morpholino)methylene]-1H-[1,2,3]triazolo[4,5-b]pyridine-1-ium 3-oxide hexafluorophosphate, N-methyl-N-[4-morpholinyl(3-oxido-1H-1,2,3-triazolo[4,5-b]pyridin-1-yl)methylene]methanaminium, hexafluorophosphate
About This Item
Recommended Products
Assay
99%
form
powder
mp
218 °C (decomposition)
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
CN(C)[C+](N1CCOCC1)N2N=[N+]([O-])C3=C2C=CC=N3.F[P-](F)(F)(F)(F)F
InChI
1S/C12H17N6O2.F6P/c1-15(2)12(16-6-8-20-9-7-16)17-10-4-3-5-13-11(10)18(19)14-17;1-7(2,3,4,5)6/h3-5H,6-9H2,1-2H3;/q+1;-1
InChI key
ILBQIVNHGIVQOQ-UHFFFAOYSA-N
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service