Skip to Content
Merck
All Photos(1)

Key Documents

774995

Sigma-Aldrich

N2-[(1,1-Dimethylethoxy)carbonyl]-N6-[(2-propynyloxy)carbonyl]-L-lysine

90%

Synonym(s):

N-α-Boc-propargyl-lysine-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O6
CAS Number:
Molecular Weight:
328.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
90%

Assay

90%

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.475

density

1.167 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC#C)C(O)=O

InChI

1S/C15H24N2O6/c1-5-10-22-13(20)16-9-7-6-8-11(12(18)19)17-14(21)23-15(2,3)4/h1,11H,6-10H2,2-4H3,(H,16,20)(H,17,21)(H,18,19)/t11-/m0/s1

InChI key

VHCJWKDEXGPNCX-NSHDSACASA-N

Application

Alkyne modified lysine that has been shown to be incorporated site-specifically into recombinant proteins.[1][2]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

164.0 °F

Flash Point(C)

73.33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of threefold glycosylated proteins using click chemistry and genetically encoded unnatural amino acids.
Emine Kaya et al.
Chembiochem : a European journal of chemical biology, 10(18), 2858-2861 (2009-11-10)
Duy P Nguyen et al.
Journal of the American Chemical Society, 131(25), 8720-8721 (2009-06-12)
We demonstrate that an orthogonal Methanosarcina barkeri MS pyrrolysyl-tRNA synthetase/tRNA(CUA) pair directs the efficient, site-specific incorporation of N6-[(2-propynyloxy)carbonyl]-L-lysine, containing a carbon-carbon triple bond, and N6-[(2-azidoethoxy)carbonyl]-L-lysine, containing an azido group, into recombinant proteins in Escherichia coli. Proteins containing the alkyne functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service