Skip to Content
Merck
All Photos(1)

Documents

773727

[2-[2-(2-Methoxyethoxy)ethoxy]ethyl] phosphonic acid ethyl ester

99% (GC)

Synonym(s):

{2-[2-(2-Methoxyethoxy)ethoxy]ethyl}phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H25O6P
CAS Number:
Molecular Weight:
284.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (GC)

form

liquid

refractive index

n20/D 1.441

density

1.087 g/mL at 25 °C

SMILES string

CCOP(=O)(CCOCCOCCOC)OCC

InChI

1S/C11H25O6P/c1-4-16-18(12,17-5-2)11-10-15-9-8-14-7-6-13-3/h4-11H2,1-3H3

InChI key

DTJZSPGSMWONOZ-UHFFFAOYSA-N

Application

The corresponding phosphoric acid after deprotection is used to modify barium titanate and related metal oxide surface leading to high permittivity values and a high dielectric strength material.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphonic acid-modified barium titanate polymer nanocomposites with high permittivity and dielectric strength
Advanced Materials, 19, 1001-1005 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service