Skip to Content
Merck
All Photos(1)

Key Documents

767611

Sigma-Aldrich

2,6-Diphenylbenzo[1,2-b:4,5-b′]dithiophene

sublimed grade, 97%

Synonym(s):

DPh-BDT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14S2
CAS Number:
Molecular Weight:
342.48
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Assay

97%

form

powder or crystals

mp

421-426 °C

semiconductor properties

P-type (mobility=4.6×10−3 cm2/V·s)

SMILES string

c1ccc(cc1)-c2cc3cc4sc(cc4cc3s2)-c5ccccc5

InChI

1S/C22H14S2/c1-3-7-15(8-4-1)19-11-17-13-22-18(14-21(17)23-19)12-20(24-22)16-9-5-2-6-10-16/h1-14H

InChI key

WNNUVWFCGFUJFU-UHFFFAOYSA-N

Application

Organic Field Effect Transistor (OFET) Materials; p-Type Organic Semiconductors; p-Type Small Molecules; sublimed grade materials

Legal Information

Product of Nippon Kayaku

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuo Takimiya et al.
Journal of the American Chemical Society, 126(16), 5084-5085 (2004-04-22)
2,6-Diphenylbenzo[1,2-b:4,5-b']dichalcogenophenes including thiophene, selenophene, and tellurophene analogues as organic semiconductors for field-effect transistors were effectively synthesized in three steps from commercially available 1,4-dibromobenzene. All three benzodichalcogenophenes acted as good p-type semiconductors, and particularly the selenophene analogue, 2,6-diphenylbenzo[1,2-b:4,5-b']diselenophene, showed high FET
Juan Casado et al.
The journal of physical chemistry. A, 110(23), 7422-7430 (2006-06-09)
In this work, the interactions between heteroatoms (S, Se, and Te) and conjugated skeletons are analyzed. The study is carried out by using electronic absorption and fluorescence spectroscopies, electrochemistry, vibrational Raman spectroscopy, and theoretical calculations in the framework of DFT

Articles

Sublimed materials for organic electronic devices such of OFETs and OTFTs allow the achievement of better electronic properties, and may help increase a device’s lifetime.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service