Skip to Content
Merck
All Photos(1)

Documents

698423

Sigma-Aldrich

Pentacene

triple-sublimed grade, ≥99.995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912418
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.995% trace metals basis

form

powder

quality

triple-sublimed grade

mp

372-374 °C (subl.)

solubility

organic solvents: slightly soluble(lit.)

Orbital energy

HOMO 5 eV 
LUMO 3 eV 

OPV Device Performance

ITO/pentacene/C60/BCP/Al

  • Short-circuit current density (Jsc): 15 mA/cm2
  • Open-circuit voltage (Voc): 0.36 V
  • Fill Factor (FF): 0.5
  • Power Conversion Efficiency (PCE): 2.7 %

semiconductor properties

P-type (mobility=0.4-3 cm2/V·s)

SMILES string

c1ccc2cc3cc4cc5ccccc5cc4cc3cc2c1

InChI

1S/C22H14/c1-2-6-16-10-20-14-22-12-18-8-4-3-7-17(18)11-21(22)13-19(20)9-15(16)5-1/h1-14H

InChI key

SLIUAWYAILUBJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Pentacene is a conjugating material with relatively high field effect mobility and ordered film-forming ability. It forms organic thin films that can be coated on a variety of substrates for the development of semiconducting devices.
TGA/DSC Lot specific traces available upon request

Application

Pentacene can be used as an active material for various optoelectronic and electronic applications such as organic light emitting diodes (OLEDs), organic thin film transistors (OTFTs) and organic photovoltaic cells (OPVs).
p-Type organic conductor. Triple-sublimed grade for organic electronic device applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentacene thin film growth
Ruiz R, et al.
Chemistry of Materials, 16(23), 4497-4508 (2004)
High-mobility polymer gate dielectric pentacene thin film transistors
Klauk H, et al.
Journal of Applied Physics, 92(9), 5259-5263 (2002)
David G Bossanyi et al.
Nature chemistry, 13(2), 163-171 (2020-12-09)
Singlet fission and triplet-triplet annihilation represent two highly promising ways of increasing the efficiency of photovoltaic devices. Both processes are believed to be mediated by a biexcitonic triplet-pair state, 1(TT). Recently however, there has been debate over the role of
Growth dynamics of pentacene thin films
Zu Heringdorf FM, et al.
Nature, 412(6846), 517-517 (2001)
Postfabrication annealing of pentacene-based photovoltaic cells
Mayer AC, et al.
Applied Physics Letters, 85(25), 6272-6274 (2004)

Articles

Flexible electronic circuits and displays based on organic active materials are future generations of products that may eventually enter mainstream electronics market.

Highly reducing or oxidizing species enhance organic semiconductor conductivity by reducing charge-carrier injection barriers.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

See All

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service