Skip to Content
Merck
All Photos(1)

Key Documents

76575

Sigma-Aldrich

1-Pentadecyne

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡C(CH2)12CH3
CAS Number:
Molecular Weight:
208.38
Beilstein:
1751009
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
≥97.0%

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.442

density

0.794 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCC#C

InChI

1S/C15H28/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h1H,4-15H2,2H3

InChI key

DONJGKADZJEXRJ-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service