Skip to Content
Merck
All Photos(2)

Key Documents

764639

Sigma-Aldrich

5,5′′′-Bis(tridecafluorohexyl)-2,2′:5′,2 ′′:5′′,2′′′-quaterthiophene

Synonym(s):

α,ω-Diperfluorohexyl-quarterthiophene, DFH-4T

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$483.00

$483.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$483.00

About This Item

Empirical Formula (Hill Notation):
C28H8F26S4
CAS Number:
Molecular Weight:
966.58
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$483.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

form

solid

Quality Level

mp

205-210 °C

semiconductor properties

N-type (mobility≤0.64 cm2/V·s)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)c1ccc(s1)-c2ccc(s2)-c3ccc(s3)-c4ccc(s4)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C28H8F26S4/c29-17(30,19(33,34)21(37,38)23(41,42)25(45,46)27(49,50)51)15-7-5-13(57-15)11-3-1-9(55-11)10-2-4-12(56-10)14-6-8-16(58-14)18(31,32)20(35,36)22(39,40)24(43,44)26(47,48)28(52,53)54/h1-8H

InChI key

UBMTYFFPSPVBSP-UHFFFAOYSA-N

General description

5,5′′′-Bis(tridecafluorohexyl)-2,2′:5′,2′′:5′′,2′′′-quaterthiophene (DFH-4T) is a n-type semiconducting material. It is a quaterthiophene derivative that contains fluoroalkyl groups and can be used as an electron transporting layer. It can be used in organic electronics.[1][2]

Application

This molecule has shown high mobilities (electron mobilities of up to 0.64 cm2/Vs) when used as an n-type semiconducting material in OFET devices.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overlapping-Gate Organic Light-Emitting Transistors
Lee J, et al.
Advanced Electronic Materials, 5(1), 1800437-1800437 (2019)
A new method of using supercritical carbon dioxide as a green solvent for synthesis and purification of 5, 5?-bis (tridecafluorohexyl)-2, 2?: 5?, 2 ?: 5 ?, 2?-quaterthiophene, which is one of n-type organic semiconducting materials
Hirase R, et al.
Tetrahedron Letters, 59(5), 469-472 (2018)
Hakan Usta et al.
Accounts of chemical research, 44(7), 501-510 (2011-05-28)
Organic semiconductors have unique properties compared to traditional inorganic materials such as amorphous or crystalline silicon. Some important advantages include their adaptability to low-temperature processing on flexible substrates, low cost, amenability to high-speed fabrication, and tunable electronic properties. These features

Articles

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service