Skip to Content
Merck
All Photos(1)

Key Documents

764221

Sigma-Aldrich

Propargyl-N-hydroxysuccinimidyl ester

Synonym(s):

2,5-Dioxopyrrolidin-1-yl 3-(prop-2-ynyloxy)propanoate, 3-(2-Propyn-1-yloxy)propanoic acid 2,5-dioxo-1-pyrrolidinyl ester, Alkyne-NHS ester, Propargyl-succinimidyl-ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$118.00
250 MG
$366.00

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$118.00
250 MG
$366.00

About This Item

Empirical Formula (Hill Notation):
C10H11NO5
CAS Number:
Molecular Weight:
225.20
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

mp

36-41 °C

functional group

NHS ester
alkyne

storage temp.

−20°C

SMILES string

C#CCOCCC(ON1C(CCC1=O)=O)=O

InChI

1S/C10H11NO5/c1-2-6-15-7-5-10(14)16-11-8(12)3-4-9(11)13/h1H,3-7H2

InChI key

WKIKHHMUNOVQLD-UHFFFAOYSA-N

Application

Propargyl-N-hydroxysuccinimidyl ester may be used as a tool for identifying nucleophilic ligandable hotspots by chemoproteomic approaches.[1]
Propargyl-NHS ester is an amine reactive reagent for derivatizing peptides, antibodies, amine coated surfaces etc, with a terminal alkyne. The alkyne can be reacted with an azide containing compound or biomolecule via copper catalyzed azide-alkyne click chemistry to yield a stable triazole linkage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activatable T1 Relaxivity Recovery Nanoconjugates for Kinetic and Sensitive Analysis of Matrix Metalloprotease
Zhu X, et al.
ACS Applied Materials & Interfaces (2017)
Qianyun Zhang et al.
ACS nano, 12(10), 10473-10485 (2018-10-06)
Multivalent presentation of ligands on nanoparticles (NPs) is considered a general strategy for enhancing receptor binding and activation through amplification of ligand-receptor interactions within the footprint of the individual NPs. The spatial clustering of ligand-functionalized NPs represents an additional, less
NHS-Esters As Versatile Reactivity-Based Probes for Mapping Proteome-Wide Ligandable Hotspots.
Ward CC, et al.
ACS Chemical Biology (2017)
Yi Yu et al.
Nature chemical biology, 16(4), 387-390 (2019-12-25)
Here, we report a rapid CRISPR-Cas9-mediated gene knock-in strategy that uses Cas9 ribonucleoprotein and 5'-modified double-stranded DNA donors with 50-base-pair homology arms and achieved unprecedented 65/40% knock-in rates for 0.7/2.5 kilobase inserts, respectively, in human embryonic kidney 293T cells. The identified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service