Skip to Content
Merck
All Photos(1)

Key Documents

763071

Sigma-Aldrich

3,5-Bis(2-dodecylthiocarbonothioylthio-1-oxopropoxy)benzoic acid

98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H62O6S6
Molecular Weight:
819.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (HPLC)

form

powder or crystals

mp

60-65 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCSC(=S)SC(C)C(=O)Oc1cc(OC(=O)C(C)SC(=S)SCCCCCCCCCCCC)cc(c1)C(O)=O

InChI

1S/C39H62O6S6/c1-5-7-9-11-13-15-17-19-21-23-25-48-38(46)50-30(3)36(42)44-33-27-32(35(40)41)28-34(29-33)45-37(43)31(4)51-39(47)49-26-24-22-20-18-16-14-12-10-8-6-2/h27-31H,5-26H2,1-4H3,(H,40,41)

InChI key

GOLRGLNMILBFFI-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.
Reversible addition-fragmentation chain transfer (RAFT) polymerization is used to produce narrow polydispersity polymers of complex architecture. 3,5-Bis(2-dodecylthiocarbonothioylthio-1-oxopropoxy)benzoic acid is a carboxylic acid functionalised trithiocarbonate RAFT agent. It is used to form well-defined block copolymers. They have good control of polymerisation for a wide range of vinyl monomers.

Application

Used in the synthesis of macro-Chain Transfer Agent (CTA) which are used in the RAFT polymerisation of vinylic monomers like styrene, acrylate and acrylamide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Block copolymers of poly (2-oxazoline) s and poly (meth) acrylates: a crossover between cationic ring-opening polymerization (CROP) and reversible addition-fragmentation chain transfer (RAFT).
Krieg, A., Weber, C., Hoogenboom, R., Becer, C. R., & Schubert, U. S.
ACS Macro Letters, 1(6), 776-779 (2012)
Thiocarbonylthio compounds (SC (Z) SR) in free radical polymerization with reversible addition-fragmentation chain transfer (RAFT polymerization).
Chiefari, J., Mayadunne, R. T., Moad, C. L., Moad, G., Rizzardo, E., Postma, A., & Thang, S. H.
Macromolecules, 36(7), 2273-2283 (2003)
One-step synthesis of block copolymers using a hydroxyl-functionalized trithiocarbonate RAFT agent as a dual initiator for RAFT polymerization and ROP
Kang, H. U., Yu, Y. C., Shin, S. J., & Youk, J. H.
Journal of Polymer Science Part A: Polymer Chemistry, 51(4), 774-779 (2013)
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service