Skip to Content
Merck
All Photos(1)

Key Documents

00505

Sigma-Aldrich

Methyl acetoacetate sodium salt

≥97.0% (T)

Synonym(s):

Sodium methyl acetoacetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$64.00

$64.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$64.00

About This Item

Linear Formula:
CH3COCHNaCOOCH3
CAS Number:
Molecular Weight:
138.10
Beilstein:
4127717
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$64.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (T)

form

powder

impurities

~0.5% water

functional group

ester

storage temp.

2-8°C

SMILES string

[Na+].COC(=O)\C=C(\C)[O-]

InChI

1S/C5H8O3.Na/c1-4(6)3-5(7)8-2;/h3,6H,1-2H3;/q;+1/p-1/b4-3-;

InChI key

PZPFXRQPPDCUQY-LNKPDPKZSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service