495557
Tetra-2-pyridinylpyrazine
97%
Synonym(s):
2,3,5,6-Tetra(2-pyridyl)pyrazine, 2,3,5,6-Tetra(2′-pyridyl)pyrazine, 2,3,5,6-Tetra-2-pyridinylpyrazine, 2,3,5,6-Tetrakis(α-pyridyl)pyrazine, 2,3,5,6-Tetrakis(2-pyridinyl)pyrazine, 2,3,5,6-Tetrakis(2-pyridyl)pyrazine, 2,3,5,6-Tetrakis(2′-pyridyl)pyrazine, Tetra(2-pyridyl)pyrazine, Tetra-2′-pyridylpyrazine
About This Item
Recommended Products
Assay
97%
mp
292-294 °C (lit.)
SMILES string
c1ccc(nc1)-c2nc(-c3ccccn3)c(nc2-c4ccccn4)-c5ccccn5
InChI
1S/C24H16N6/c1-5-13-25-17(9-1)21-22(18-10-2-6-14-26-18)30-24(20-12-4-8-16-28-20)23(29-21)19-11-3-7-15-27-19/h1-16H
InChI key
UOJZYBFRNITHCX-UHFFFAOYSA-N
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service