Skip to Content
Merck
All Photos(2)

Key Documents

756512

Sigma-Aldrich

Barium magnesium aluminate, europium doped

99% trace metals basis

Synonym(s):

Aluminum barium europium magnesium oxide, Blue phosphor, Europium doped aluminum barium magnesium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ba0.86Eu0.14MgAl10O17
CAS Number:
Molecular Weight:
705.49
UNSPSC Code:
12352300
NACRES:
NA.23

Assay

99% trace metals basis

form

powder

composition

Ba0.86Eu0.14MgAl10O17

particle size

5-9 μm (d50)

mp

>1300 °C (lit.)

fluorescence

λem 450 nm±10 nm (UV excitation source)

General description

Barium magnesium aluminate, europium doped is an efficient phosphor that is majorly used as a light emitter. It can be prepared by low temperature combustion process by utilizing urea as a fuel. It shows a band gap of ~5.24 eV and a crystallite size in the order of ~70 nm−650 nm. The thermo-luminescence (TL) spectra potentially peak at ~443 nm.[1][2]

Application

Barium magnesium aluminate, europium doped can be used in the fabrication of new hybrid perovskites and light emitting diodes (LEDs).[1][2][3]

Other Notes

Color coordinate, x: 0.140 +/- 0.015
Color coordinate, y: 0.050 +/- 0.010

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunimi, S.; Fujihara, S.
Dyes and Pigments, 91, 49-49 (2011)
Taden, A.; et al.
Chemistry of Materials, 16, 5081-5081 (2004)
Yoshida, J.; et al.
Journal of the Electrochemical Society, 154, J196-J196 (2007)
Full Spectrum White LEDs of Any Color Temperature with Color Rendering Index Higher Than 90 Using a Single Broad-Band Phosphor
Dutta PS and Liotta KM
ECS journal of solid state science and technology : JSS, 7(1), R3194-R3198 (2018)
Screening Approach for the Discovery of New Hybrid Perovskites with Efficient Photoemission
Brochard-Garnier S, et al.
Advances in Functional Materials, 29(1), 1806728-1806728 (2019)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service