Skip to Content
Merck
All Photos(1)

Key Documents

754102

Sigma-Aldrich

TES-ADT

>99% (HPLC)

Synonym(s):

5,11-Bis(triethylsilylethynyl)anthradithiophene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$460.00

$460.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$460.00

About This Item

Empirical Formula (Hill Notation):
C34H38S2Si2
CAS Number:
Molecular Weight:
566.97
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$460.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

>99% (HPLC)

form

solid

loss

0.5 wt. % TGA, 250 °C

mp

128-135 °C

semiconductor properties

P-type (mobility=1 cm2/V·s)

SMILES string

CC[Si](CC)(CC)C#Cc1c2cc3ccsc3cc2c(C#C[Si](CC)(CC)CC)c4cc5ccsc5cc14

InChI

1S/C34H38S2Si2/c1-7-37(8-2,9-3)19-15-27-29-21-25-13-17-36-34(25)24-32(29)28(16-20-38(10-4,11-5)12-6)30-22-26-14-18-35-33(26)23-31(27)30/h13-14,17-18,21-24H,7-12H2,1-6H3

InChI key

NYBWUHOMYZZKOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service