Skip to Content
Merck
All Photos(1)

Documents

752142

Sigma-Aldrich

n-Butyl-sec-butylmagnesium solution

0.7 M in hexane

Synonym(s):

Butyl(1-methylpropyl)magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18Mg
Molecular Weight:
138.53
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

solution

concentration

0.7 M in hexane

density

0.695 g/mL at 25 °C

SMILES string

CCCC[Mg]C(C)CC

InChI

1S/2C4H9.Mg/c2*1-3-4-2;/h3H,4H2,1-2H3;1,3-4H2,2H3;

InChI key

DBNGNEQZXBWPTJ-UHFFFAOYSA-N

Application

n-Butyl-sec-butylmagnesium can be used as a starting material in the synthesis of magnesium bis(alkoxide) complexes applicable in the ring-opening polymerization and co-polymerization reactions.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

<-22.0 °F

Flash Point(C)

< -30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a mononuclear magnesium bis(alkoxide) complex and its reactivity in the ring-opening copolymerization of cyclic anhydrides with epoxides
Wannipurage D, et al.
Dalton Transactions, 49(8), 2715-2723 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service