Skip to Content
Merck
All Photos(1)

Key Documents

291005

Sigma-Aldrich

Butylmagnesium chloride solution

2.0 M in THF

Synonym(s):

BuMgCl

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$124.00
800 ML
$317.00

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$124.00
800 ML
$317.00

About This Item

Linear Formula:
CH3(CH2)3MgCl
CAS Number:
Molecular Weight:
116.87
Beilstein:
3587228
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in THF

density

0.962 g/mL at 25 °C

SMILES string

CCCC[Mg]Cl

InChI

1S/C4H9.ClH.Mg/c1-3-4-2;;/h1,3-4H2,2H3;1H;/q;;+1/p-1

InChI key

UUNGXYVLXYGZKH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Butylmagnesium chloride (BuMgCl) is a Grignard reagent that can be used:
  • To prevent the formation of passivating surface films on Mg electrodes.[1]
  • For the synthesis of lithium tributylmagnesate complex (n-Bu3MgLi) by reacting with n-BuLi.[2]

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tributylmagnesium ate complex-mediated novel bromine-magnesium exchange reaction for selective monosubstitution of dibromoarenes.
Iida T, et al.
Tetrahedron Letters, 42(29), 4841-4844 (2001)
On the electrochemical behavior of magnesium electrodes in polar aprotic electrolyte solutions.
Lu Z, et al.
Journal of Electroanalytical Chemistry, 466(2), 203-217 (1999)
B Pons et al.
Journal of chromatography. B, Biomedical sciences and applications, 716(1-2), 139-145 (1998-11-21)
One analytical procedure for the determination of ionic alkyllead in human urine has been studied. The system consists of the extraction of Me3Pb+, Et3Pb+ and Pb2+ at pH 9.0 with diethyldithiocarbamate to an organic phase. Then, the ionic compounds are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service