Skip to Content
Merck
All Photos(1)

Key Documents

746622

Sigma-Aldrich

SAM Formation reagent for biotin terminated monolayer on gold surface

Synonym(s):

Biotin-SAM

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$745.00

$745.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
$745.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

$745.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

storage temp.

2-8°C

Application

Because the Biotin-Avidin formation is known as a facile reaction, this product is widely used to immobilize a protein on a Surface Plasmon Resonance (SPR) or Quartz Crystal Microbalance (QCM) biosensor consisting of a Self-Assembled Monolayer (SAM).

Used for:
  • Simple preparation of Biotin-SAM on gold surface
  • Efficient Immobilization of Avidins
  • Suppress unspecific binding with unknown proteins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiko Ishizuka-Katsura et al.
Journal of bioscience and bioengineering, 105(5), 527-535 (2008-06-19)
We describe a technique to form a biotin-containing phospholipid vesicle layer on a self-assembled monolayer (SAM) deposited on a gold surface to immobilize biotinylated receptor proteins for a surface plasmon resonance (SPR) biosensor. The adsorption of vesicle of 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC)
Jennifer S Shumaker-Parry et al.
Analytical chemistry, 76(4), 918-929 (2004-02-14)
We present two strategies for microspotting 10 x 12 arrays of double-stranded DNAs (dsDNAs) onto a gold-coated glass slide for high-throughput studies of protein-DNA interactions by surface plasmon resonance (SPR) microscopy. Both methods use streptavidin (SA) as a linker layer

Questions

  1. What is the nature of the SAM used in the 746622 SAM-Biotin product when applied to Au surfaces? Specifically, is the SAM type proprietary, and could information on the length of C atoms in alkanethiol or the presence of any functional groups like -COOH be provided, if present on the SAM other than Biotin?

    1 answer
    1. The chemical structure and SAM type for this product are not disclosed

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service