Skip to Content
Merck
All Photos(3)

Key Documents

741248

Sigma-Aldrich

Bis(triphenylphosphine)palladium(II) dichloride

Synonym(s):

Dichlorobis(triphenylphosphine)palladium(II), Palladium(II)bis(triphenylphosphine) dichloride, PdCl2(PPh3)2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$86.20
5 G
$327.00
25 G
$629.00

$86.20


Estimated to ship onApril 09, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$86.20
5 G
$327.00
25 G
$629.00

About This Item

Linear Formula:
[(C6H5)3P]2PdCl2
CAS Number:
Molecular Weight:
701.90
Beilstein:
4935975
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$86.20


Estimated to ship onApril 09, 2025


Request a Bulk Order

form

solid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
core: palladium

reagent type: ligand

functional group

phosphine

SMILES string

Cl[Pd]Cl.c1(P(c2ccccc2)c3ccccc3)ccccc1.c4(P(c5ccccc5)c6ccccc6)ccccc4

InChI

1S/2C18H15P.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

InChI key

YNHIGQDRGKUECZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Bis(triphenylphosphine)palladium(II) dichloride is generally used as a catalyst for the synthesis of conjugated polymers via Stille coupling.[1] It is also applicable to other coupling reactions such as Suzuki-[2] and Sonogashira-coupling reactions[3]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of quaternized zinc (II) and cobalt (II) phthalocyanines bearing pyridine-2-yl-ethynyl groups and their DNA binding properties.
Alishah MM, et al.
Turkish Journal of Chemistry, 42(2), 572-585 (2018)
New oligomeric poly (ether-imide) s containing diphenylsilane and dibenzofuran moieties. Synthesis and characterization.
Tundidor-Camba A, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 55(4), 324-331 (2018)
Influence of catalytic systems in Stille polymerization on the electrochromic performance of diketopyrrolopyrrole-based conjugated polymers.
Neo WT, et al.
Materials Chemistry Frontiers. (2018)
Ganesan Bharathiraja et al.
Organic & biomolecular chemistry, 13(9), 2786-2792 (2015-01-22)
Various tetrasubstituted pyrroles/pyrazoles have been prepared from nitro-substituted 1,3-enynes with aromatic amines/hydrazines via a copper-catalyzed cascade aza-Michael addition, cyclization and aromatization at room temperature. This protocol is also effective for the synthesis of tetrasubstituted pyrazoles in high yields.
Hiroaki Imoto et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12105-12111 (2015-07-17)
Aggregation-induced emission (AIE)-active maleimide dyes, namely, 2-p-toluidino-N-p-tolylmaleimide, 3-phenyl-2-toluidino-N-p-tolylmaleimide, 2-p-thiocresyl-3-p-toluidino-N-p-tolylmaleimide, and 2,3-dithiocresyl-N-arylmaleimides, were synthesized by facile synthetic procedures. The dyes show intense emission in the solid state, and emission colors were controlled from green (λmax =527 nm) to orange (λmax =609 nm) by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service