Skip to Content
Merck
All Photos(1)

Key Documents

720356

Sigma-Aldrich

3-Phenyl-2-propyn-1-amine hydrochloride

97%

Synonym(s):

3-Phenylpropargylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$199.00

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$199.00

About This Item

Empirical Formula (Hill Notation):
C9H10ClN
CAS Number:
Molecular Weight:
167.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

222-226 °C

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

Cl.NCC#Cc1ccccc1

InChI

1S/C9H9N.ClH/c10-8-4-7-9-5-2-1-3-6-9;/h1-3,5-6H,8,10H2;1H

InChI key

NOXMJZCFMAEDRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dopamine β-hydroxylase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service