Skip to Content
Merck
All Photos(4)

Documents

740845

Sigma-Aldrich

5-[Di(1-adamantyl)phosphino]-1′,3′,5′-triphenyl-1′H-[1,4′]bipyrazole

97%

Synonym(s):

Ad-BGPhos, 5-[Bis(1-adamantyl)phosphino]-1′,3′,5′-triphenyl-1,4′-bi-1H-pyrazole, Adamantyl-BippyPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H47N4P
CAS Number:
Molecular Weight:
662.84
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

295-300 °C

storage temp.

2-8°C

SMILES string

C1[C@@H]2C[C@@H]3C[C@H]1C[C@](C2)(C3)P(c4ccnn4-c5c(nn(-c6ccccc6)c5-c7ccccc7)-c8ccccc8)[C@@]9%10C[C@@H]%11C[C@@H](C[C@@H](C%11)C9)C%10

InChI

1S/C44H47N4P/c1-4-10-36(11-5-1)40-42(41(37-12-6-2-7-13-37)47(46-40)38-14-8-3-9-15-38)48-39(16-17-45-48)49(43-24-30-18-31(25-43)20-32(19-30)26-43)44-27-33-21-34(28-44)23-35(22-33)29-44/h1-17,30-35H,18-29H2/t30-,31+,32-,33-,34+,35-,43-,44-

InChI key

IHFBOUNFWVRWAQ-GOSCEEDCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-[Di(1-adamantyl)phosphino]-1′,3′,5′-triphenyl-1′H-[1,4′]bipyrazole (Ad-BippyPhos) is an effective ligand that catalyzes C-O coupling reactions of aryl and heteroaryl bromides/chlorides with primary aliphatic alcohols. It can also be used as a catalyst in the synthesis of fluorinated anilines by coupling fluoroalkylamines with aryl bromides and chlorides.

Other Notes

Store at 2-8 °C.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Gowrisankar, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 2498-2502 (2012)
Palladium-catalyzed arylation of fluoroalkylamines.
Brusoe A T and Hartwig J F
Journal of the American Chemical Society, 137(26), 8460-8468 (2015)
A general and efficient catalyst for palladium-catalyzed C? O coupling reactions of aryl halides with primary alcohols.
Gowrisankar S, et al.
Journal of the American Chemical Society, 132(33), 11592-11598 (2010)
Palladium-Catalyzed C?O Coupling of a Sterically Hindered Secondary Alcohol with an Aryl Bromide and Significant Purity Upgrade in the API Step.
Young I S, et al.
Organic Process Research & Development, 22(5), 585-594 (2018)
Gowrisankar, S.; et al.
ChemCatChem, 3, 1439-1441 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service