Skip to Content
Merck
All Photos(1)

Documents

739898

Sigma-Aldrich

1α,25-Dihydroxyvitamin D2 solution

50 μg/mL in ethanol, 95% (CP)

Synonym(s):

1α,25-Dihydroxycalciferol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O3
CAS Number:
Molecular Weight:
428.65
EC Number:
UNSPSC Code:
12352205
NACRES:
NA.24

Quality Level

Assay

95% (CP)

form

solution

concentration

50 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

storage temp.

−20°C

SMILES string

CC(O)(C)[C@@H](C)/C=C/[C@@H](C)[C@H]1CCC([C@]1(C)CCC/2)C2=C\C=C(C[C@@H](O)C[C@@H]3O)/C3=C

InChI

1S/C28H44O3/c1-18(9-10-19(2)27(4,5)31)24-13-14-25-21(8-7-15-28(24,25)6)11-12-22-16-23(29)17-26(30)20(22)3/h9-12,18-19,23-26,29-31H,3,7-8,13-17H2,1-2,4-6H3/b10-9+,21-11+,22-12-/t18-,19+,23-,24-,25?,26+,28-/m1/s1

InChI key

ZGLHBRQAEXKACO-KPKHAADHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1α,25-Dihydroxyvitamin D2 is an active form of vitamin D2. It is also known as ercalcitriol or 1α,25-dihydroxyergocalciferol. It was first isolated from mitochondria of chick kidney.

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convergent synthesis of double point modified analogs of 1α, 25-dihydroxyvitamin D2 for biological evaluation
Nadkarni S, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 164, 45-49 (2016)
Marc Lamblin et al.
Journal of medicinal chemistry, 53(20), 7461-7465 (2010-10-05)
Vitamin D receptor (VDR) antagonists have therapeutic potential in treatment of allergic conditions and hypercalcemia driven by granulomatous diseases. We have identified an o-aminoanilide analogue of the hormonal form of vitamin D with a dienyl side chain that functions as
Pierre Antony et al.
Journal of medicinal chemistry, 53(3), 1159-1171 (2010-01-15)
The vitamin D nuclear receptor is a ligand-dependent transcription factor that controls multiple biological responses such as cell proliferation, immune responses, and bone mineralization. Numerous 1 alpha,25(OH)(2)D(3) analogues, which exhibit low calcemic side effects and/or antitumoral properties, have been synthesized.
Angsuma Wongmayura et al.
Bioorganic & medicinal chemistry letters, 22(4), 1756-1760 (2012-01-31)
Vitamin D receptor (VDR) is a nuclear receptor for 1α,25-dihydroxyvitamin D(3) (1α,25(OH)(2)D(3)), and is an attractive target for multiple clinical applications. We recently developed novel non-secosteroidal VDR ligands bearing a hydrophobic p-carborane cage, thereby establishing the utility of this spherical
Yuka Inaba et al.
Journal of medicinal chemistry, 52(5), 1438-1449 (2009-02-06)
To identify novel vitamin D receptor (VDR) ligands that induce a novel architecture within the ligand-binding pocket (LBP), we have investigated eight 22-butyl-1alpha,24-dihydroxyvitamin D(3) derivatives (3-10), all having a butyl group as the branched alkyl side chain. We found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service