Skip to Content
Merck
All Photos(1)

Documents

740500

Sigma-Aldrich

Chitosan

high purity, Mv 110,000-150,000

Synonym(s):

KiOmedine-CsU® C, Deacetylated chitin, Poly(D-glucosamine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201

biological source

white mushroom

grade

high purity

form

powder

mol wt

Mv 110,000-150,000

composition

Bacterial endotoxins, ≤10 EU/g
Beta-glucan, ≤5 % (w/w)
Chitosan, ≥93 % (w/w)
Proteins, ≤1 % (w/w)

viscosity

5-200 mPa.s (1% solution in 1% HAc)

solubility

acetic acid: 1% (solution)

storage temp.

2-8°C

InChI

1S/C56H103N9O39/c1-87-56(86)65-28-38(84)46(19(10-74)96-55(28)104-45-18(9-73)95-49(27(64)37(45)83)97-39-12(3-67)88-47(85)20(57)31(39)77)103-54-26(63)36(82)44(17(8-72)94-54)102-53-25(62)35(81)43(16(7-71)93-53)101-52-24(61)34(80)42(15(6-70)92-52)100-51-23(60)33(79)41(14(5-69)91-51)99-50-22(59)32(78)40(13(4-68)90-50)98-48-21(58)30(76)29(75)11(2-66)89-48/h11-55,66-85H,2-10,57-64H2,1H3,(H,65,86)/t11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48+,49+,50+,51+,52+,53+,54+,55+/m1/s1

InChI key

FLASNYPZGWUPSU-SICDJOISSA-N

Looking for similar products? Visit Product Comparison Guide

General description

KiOmedine-CSU® is an ultra-pure chitosan of non-animal origin, produced from white mushrooms (Agaricus bisporus), in accordance with cGMP.
The crystalline structure is in the alpha form. The linkage between glucosamine units is beta(1,4).

Application

Forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films.
Ideal for wound healing and hemostasis; biosurgery and ophthalmology, scaffold and cell therapy; and drug delivery and vaccines

Features and Benefits

Biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.

Physical properties

Degree of acetylation: ≤40 mol. %

Legal Information

Product of KitoZyme
KiOmedine is a registered trademark of KitoZyme, S.A.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alberto Busilacchi et al.
Carbohydrate polymers, 98(1), 665-676 (2013-08-31)
The idea of using chitosan as a functional delivery aid to support simultaneously PRP, stem cells and growth factors (GF) is associated with the intention to use morphogenic biomaterials to modulate the natural healing sequence in bone and other tissues.
Stephanie Supper et al.
Expert opinion on drug delivery, 11(2), 249-267 (2013-12-07)
Thermogelling chitosan (CS)/glycerophosphate (GP) solutions have been reported as a new type of parenteral in situ forming depot system. These free-flowing solutions at ambient temperature turn into semi-solid hydrogels after parenteral administration. Formulation parameters such as CS physico-chemical characteristics, CS/gelling
Liming Hu et al.
Nanoscale, 5(8), 3103-3111 (2013-03-22)
Within the past few years, chitosan-based drug delivery vehicles have become some of the most attractive to be studied. In contrast to all other polysaccharides, chitosan has demonstrated its unique characteristics for drug delivery platforms, including its active primary amino
William D Stanish et al.
The Journal of bone and joint surgery. American volume, 95(18), 1640-1650 (2013-09-21)
Microfracture, the standard of care, is recognized to be an incomplete solution for cartilage damage. BST-CarGel, a chitosan-based medical device, is mixed with autologous whole blood and is applied to a microfractured cartilage lesion in which it physically stabilizes the
Electrochemical biosensor applications of polysaccharides chitin and chitosan.
Wipa Suginta et al.
Chemical reviews, 113(7), 5458-5479 (2013-04-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service