Skip to Content
Merck
All Photos(1)

Key Documents

738085

Sigma-Aldrich

Isophorone-2,4,4,6,6-d5

97 atom % D, 95% (CP)

Synonym(s):

3,5,5-Trimethyl-2-cyclohexen-1-one-2,4,4,6,6-d5

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$550.00
25 MG
$1,240.00
50 MG
$2,300.00
100 MG
$4,190.00

$550.00


Estimated to ship onMay 26, 2025


Request more information

Select a Size

Change View
10 MG
$550.00
25 MG
$1,240.00
50 MG
$2,300.00
100 MG
$4,190.00

About This Item

Empirical Formula (Hill Notation):
C9H9D5O
CAS Number:
Molecular Weight:
143.24
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.24

$550.00


Estimated to ship onMay 26, 2025


Request more information

isotopic purity

97 atom % D

Assay

95% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

bp

213-214 °C (lit.)

mp

-8 °C (lit.)

density

0.956 g/mL at 25 °C

mass shift

M+5

storage temp.

−20°C

SMILES string

[2H]C1=C(C)C([2H])([2H])C(C)(C)C([2H])([2H])C1=O

InChI

1S/C9H14O/c1-7-4-8(10)6-9(2,3)5-7/h4H,5-6H2,1-3H3/i4D,5D2,6D2

InChI key

HJOVHMDZYOCNQW-FIFBNPHPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service