Skip to Content
Merck
All Photos(1)

Documents

734322

Sigma-Aldrich

cis-5,8,11,14,17-Eicosapentaenoic acid-19,19,20,20,20-d5

98 atom % D, 98% (CP)

Synonym(s):

EPD-d5, Timnodonic acid-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20D5H25O2
CAS Number:
Molecular Weight:
307.48
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

mp

-54--53 °C (lit.)

density

0.958 g/mL at 25 °C

mass shift

M+11

storage temp.

−20°C

SMILES string

OC(CCC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C([2H])([2H])C([2H])([2H])[2H])=O

InChI

1S/C20H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-19H2,1H3,(H,21,22)/b4-3-,7-6-,10-9-,13-12-,16-15-/i1D3,2D2

InChI key

JAZBEHYOTPTENJ-YSTLBCQOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service