Skip to Content
Merck
All Photos(1)

Key Documents

73749

Sigma-Aldrich

Nα-Fmoc-Nε-biotinyl-L-lysine

≥95.0% (HPLC)

Synonym(s):

Nα-Biotinyl-Nε-Fmoc-L-lysine, Fmoc-Lys(biotinyl)-OH

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$598.00

$598.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$598.00

About This Item

Empirical Formula (Hill Notation):
C31H38N4O6S
CAS Number:
Molecular Weight:
594.72
MDL number:
UNSPSC Code:
12352209
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.26

$598.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

SMILES string

[H][C@@]12CS[C@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3c4ccccc4-c5ccccc35)C(O)=O)[C@]1([H])NC(=O)N2

InChI

1S/C31H38N4O6S/c36-27(15-6-5-14-26-28-25(18-42-26)33-30(39)35-28)32-16-8-7-13-24(29(37)38)34-31(40)41-17-23-21-11-3-1-9-19(21)20-10-2-4-12-22(20)23/h1-4,9-12,23-26,28H,5-8,13-18H2,(H,32,36)(H,34,40)(H,37,38)(H2,33,35,39)/t24-,25-,26-,28-/m0/s1

InChI key

OFIBQNGDYNGUEZ-OBXRUURASA-N

Other Notes

Synthesis of site-specific biotinylated probes[1]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Macielag et al.
International journal of peptide and protein research, 44(6), 582-588 (1994-12-01)
The solid-phase synthesis of two porcine motilin derivatives, specifically biotinylated on the side chain of Lys20, was accomplished by preactivation of the protected amino acids N alpha-(9-fluorenylmethoxycarbonyl)-N epsilon-biotinyl-L-lysine and N alpha-(9-fluorenylmethoxycarbonyl)-N epsilon-[N-(biotinyl)-6-aminohexanoyl]-L-lysine with BOP/HOBt/DIEA (1:1:2.5) followed by coupling to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service