Skip to Content
Merck

Skip To

8.52012

Fmoc-Lys(Boc)-OH

Novabiochem®

Synonym(s):

Fmoc-Lys(Boc)-OH, N-α-Fmoc-N-ε-t.-Boc-L-lysine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H32N2O6
CAS Number:
Molecular Weight:
468.54
UNSPSC Code:
12352209
EC Index Number:
276-256-4
NACRES:
NA.22
MDL number:
eCl@ss:
32160406
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

Novabiochem®

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

123-130 °C

application(s)

peptide synthesis

functional group

Boc, amine

storage temp.

2-30°C

SMILES string

N(C(CCCCNC(=O)OC(C)(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C26H32N2O6/c1-26(2,3)34-24(31)27-15-9-8-14-22(23(29)30)28-25(32)33-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H,27,31)(H,28,32)(H,29,30)

InChI key

UMRUUWFGLGNQLI-UHFFFAOYSA-N

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities. Standard building block for introduction of lysine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Application


  • Pharmacokinetics of Tizanidine in Muscle Spasticity: A comprehensive review on Tizanidine covers its pharmacological advancements and therapeutic applications, particularly focusing on muscle spasticity. This highlights the relevance of high-purity Tizanidine-D₄ hydrochloride in creating precise and reproducible pharmacokinetic profiles (Zhu et al., 2024).

  • Stable Isotope-Labeled Tizanidine for Bioanalytical Method Development: The development and validation of bioanalytical methods for monitoring drug interactions and metabolism extensively utilize Tizanidine-D₄ HCl. The isotopic labeling aids in accurate quantification and identification, enhancing the reliability of pharmacokinetic and pharmacodynamic studies (Chen et al., 2024).

  • Tizanidine-D₄ Hydrochloride Solution for Receptor Binding Studies: The application of Tizanidine-D₄ in receptor binding studies is inferred from research exploring its withdrawal effects and the pharmacological interventions required, suggesting its utility in detailed receptor interaction studies in clinical settings (Tanimura et al., 2024).


Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Lys(Boc)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Lys(Boc)-Lys(Boc)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Lys-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1026

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.521468.520138.52023
form

powder

form

powder

form

powder

form

powder

assay

≥98% (TLC), ≥99.0% (HPLC), ≥98.0% (acidimetric)

assay

≥97.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

assay

≥95.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

assay

≥97.0% (HPLC), ≥98% (TLC)

functional group

Boc, amine

functional group

amine

functional group

amine

functional group

amine

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

2-30°C

storage temp.

2-30°C

mp

123-130 °C

mp

120-130 °C

mp

105-115 °C

mp

-


Still not finding the right product?

Explore all of our products under Fmoc-Lys(Boc)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yun Song et al.
Cell reports, 30(8), 2699-2711 (2020-02-27)
The transcriptional corepressor complex CoREST is one of seven histone deacetylase complexes that regulate the genome through controlling chromatin acetylation. The CoREST complex is unique in containing both histone demethylase and deacetylase enzymes, LSD1 and HDAC1, held together by the



Global Trade Item Number

SKUGTIN
852012050004027269179588
852012500004027269221560
852012010004027269085155
852012102404061841211112
852012100004027269130466
852012200004027269232757
852012025004027269206970
852012002504027269085148

Questions

Reviews

No rating value

Active Filters