Skip to Content
Merck
All Photos(1)

Key Documents

737453

Sigma-Aldrich

N-(2-Hydroxyethyl)formamide

95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$648.00

$648.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$648.00

About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$648.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.474

density

1.160 g/mL at 25 °C

functional group

amide
hydroxyl

storage temp.

2-8°C

SMILES string

OCCNC=O

InChI

1S/C3H7NO2/c5-2-1-4-3-6/h3,5H,1-2H2,(H,4,6)

InChI key

BAMUPQJDKBGDPU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service