Skip to Content
Merck
All Photos(1)

Key Documents

576166

Sigma-Aldrich

1-(2-Hydroxyethyl)imidazole

97%

Synonym(s):

1-(β-Hydroxyethyl)imidazole, 1-(2-Hydroxyethyl)-1H-imidazole, 1-(2′-Hydroxyethyl)imidazole, 2-(1H-Imidazol-1-yl)ethan-1-ol, 2-(1H-Imidazol-1-yl)ethanol, 2-(1′-Imidazolyl)ethanol, 2-(Imidazol-1-yl)ethanol, N-(2-Hydroxyethyl)-1H-imidazole, N-(2-Hydroxyethyl)imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$84.70

$84.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$84.70

About This Item

Empirical Formula (Hill Notation):
C5H8N2O
CAS Number:
Molecular Weight:
112.13
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$84.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

36-40 °C (lit.)

functional group

hydroxyl

SMILES string

OCCn1ccnc1

InChI

1S/C5H8N2O/c8-4-3-7-2-1-6-5-7/h1-2,5,8H,3-4H2

InChI key

AMSDWLOANMAILF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yingying Ding et al.
ACS applied materials & interfaces, 11(3), 2860-2869 (2018-12-27)
Gram-negative bacteria, containing toxic proinflammatory and pyrogenic substances [endotoxin or lipopolysaccharide (LPS)], can lead to infection and associated serious diseases, such as sepsis and septic shock. Development of antimicrobial materials with intrinsically endotoxin adsorption activity can prevent the release of
Yiding Zhang et al.
Analytical and bioanalytical chemistry, 408(30), 8655-8661 (2016-08-06)
The online combination of capillary electrophoresis (CE) with mass spectrometry (MS) has long been desired for the capability of direct and simultaneous separation and detection with high efficiency, accuracy, and throughput. In this work, a novel CE-MS interface was developed
Xinxue Zhang et al.
Journal of chromatography. A, 1620, 461013-461013 (2020-03-24)
Dummy molecularly imprinted microspheres (DMIMs) were synthesized by Pickering emulsion polymerization and used as the matrix solid-phase dispersion extraction (MSPD) sorbent for sample pre-treatment of azole fungicides in fish samples. Alpha-(2,4-Dichlorophenyl)-1H-imidazole-1-ethanol (DCE) was used as the fragment dummy template for
Lorena Baranda Pellejero et al.
Nature communications, 11(1), 6242-6242 (2020-12-09)
DNA-templated synthesis takes advantage of the programmability of DNA-DNA interactions to accelerate chemical reactions under diluted conditions upon sequence-specific hybridization. While this strategy has proven advantageous for a variety of applications, including sensing and drug discovery, it has been so

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service