Skip to Content
Merck
All Photos(1)

Key Documents

735507

Sigma-Aldrich

(S)-1-Boc-3-pyrrolidinecarboxylic acid

97%

Synonym(s):

(S)-1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid, N-Boc-D-β-proline

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$181.00

$181.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$181.00

About This Item

Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$181.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

optical activity

[α]/D +15.0°, c = 0.5% in chloroform

mp

140-145 °C

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CC[C@@H](C1)C(O)=O

InChI

1S/C10H17NO4/c1-10(2,3)15-9(14)11-5-4-7(6-11)8(12)13/h7H,4-6H2,1-3H3,(H,12,13)/t7-/m0/s1

InChI key

HRMRQBJUFWFQLX-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service