Skip to Content
Merck
All Photos(1)

Key Documents

47595

Sigma-Aldrich

Fmoc-Sar-OH

≥98.0%

Synonym(s):

Fmoc-N-methylglycine, Fmoc-sarcosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$191.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$191.00

About This Item

Empirical Formula (Hill Notation):
C18H17NO4
CAS Number:
Molecular Weight:
311.33
Beilstein:
6660958
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98.0%

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CN(CC(O)=O)C(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C18H17NO4/c1-19(10-17(20)21)18(22)23-11-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,16H,10-11H2,1H3,(H,20,21)

InChI key

ZHKQIADIIYMFOZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tatsuhiko Kondo et al.
Plant & cell physiology, 52(1), 30-36 (2010-10-12)
CLAVATA3 (CLV3) is a plant peptide hormone in which the proline residues are post-translationally hydroxylated and glycosylated. CLV3 plays a key role in controlling the stem cell mass in the shoot meristem of Arabidopsis thaliana. In a previous report, we
Ewa Radzikowska et al.
Organic & biomolecular chemistry, 13(1), 269-276 (2014-11-05)
Chimeric oligonucleotides containing phosphodiester and phosphorothioate linkages have been obtained using the solid phase synthesis. The oligonucleotide parts possessing natural internucleotide phosphate bonds were assembled using commercially available nucleoside 3'-O-(2-cyanoethyl-N,N-diisopropylamino)phosphoramidites 7 whereas the phosphorothioate segment was built using nucleoside 3'-O-(2-thio-1,3,2-oxathiaphospholanes)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service